VWR® Reagent Bottles with PP 28 Thread
1000 ml, PP 28, 100,5 mm, 206,2 mm
Katalog # 215-4409
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
- Good chemical and physical properties
- Clear or amber glass
- Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Ordering information: Supplied without caps, screw caps must be ordered separately.
Specifikationer
- Udv.Ø:100,5 mm
- Gevind:PP 28
- Kapacitet:1000 ml
- Højde:206,2 mm
- Seq:07



