VWR® Reagent Bottles with PP 28 Thread
500 ml, PP 28, 82 mm, 161 mm
215-4408
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
- Good chemical and physical properties
- Clear or amber glass
- Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
: Supplied without caps, screw caps must be ordered separately.
- Udv.Ø:82 mm
- Gevind:PP 28
- Kapacitet:500 ml
- Højde:161 mm
- Seq:06