VWR® Reagent Bottles with PP 28 Thread
250 ml, PP 28, 65 mm, 133 mm
Katalog # 215-4407
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
- Good chemical and physical properties
- Clear or amber glass
- Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
: Supplied without caps, screw caps must be ordered separately.
Specifikationer
- Udv.Ø:65 mm
- Gevind:PP 28
- Kapacitet:250 ml
- Højde:133 mm
- Seq:05



