VWR® Reagent Bottles with PP 28 Thread
30 ml, PP 28, 34,2 mm, 69 mm
Katalog # 215-4403
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
- Good chemical and physical properties
- Clear or amber glass
- Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
: Supplied without caps, screw caps must be ordered separately.
Specifikationer
- Udv.Ø:34,2 mm
- Gevind:PP 28
- Kapacitet:30 ml
- Højde:69 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



