Docusate natrium USP
Poly pail, 2.500 g
Katalog # SPCMDO105-2.5KG
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonymer: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS nummer:577-11-7
- Formel:C₂₀H₃₇NaO₇S
- MDL nummer:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:12,03460
- Kogende Pt:∼180 °C (1013 hPa)
- Smeltende Pt:∼155 °C
- Tæthed:∼1,1 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-406-4
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Assay:99% Min.
Specifikationer
- Pakning:Poly pail
- Pakn:2.500 g
- Cat. No.:SPCMDO105-2.5KG