PARAPLAST REGULAR BULK PACK 56C 1 * 5 kg
LEIH39602012
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Q Path®, Biopsy Pads
: VWR Collection
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
VWR®, Nitril handsker, NITRILE
: VWR Collection
VWR® Bench Protection Papers
: VWR Collection
Clean-Lab, Q Path® for paraffin removal
: VWR Chemicals
VWR®, Ansigtsservietter
: VWR Collection
Cryobokse, 133x133 mm
: RATIOLAB
Microtome blades
: LEICA HISTOLOGY
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Ethanol Eurodenatured ≥99%, TechniSolv® (1:1:1)
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Cervical brushes, Cervex-Brush®
: Rovers