Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HighDuster covers, PREMIRA® II

HighDuster covers, PREMIRA® II

PREMIRA® II microfibre highduster covers, 79,4% polyester/20,6% polypropylene, 53,3×7,6 cm
Katalog # CONTPRMW0001
Leverandør:   CONTEC
HighDuster covers, PREMIRA® II
Katalog # CONTPRMW0001
Leverandør:  CONTEC
HighDuster covers, PREMIRA® II
HighDuster covers, PREMIRA® II

PREMIRA® II microfibre highduster covers, 79,4% polyester/20,6% polypropylene, 53,3×7,6 cm

Price
1.830,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Disposable Premira II Microfibre HighDuster covers reduce fibre shedding and provide effective dry and damp dusting for hard to reach areas. The microfibre fabric offers excellent pickup power and is compliant with most cleaning protocols. Compatible with the flexible foam snake duster.

  • Ideal for hard to reach places
  • Easily slips onto a variety of dusting frames
  • Disposable, clinically superior alternative to standard dust wands
  • Picks up and holds onto debris, rather that spreading it around
  • Conformable flexible foam Snake Duster reduces operator strain

Compatible with all QuickConnect™ handles.

Specifikationer
  • Beskrivelse:
    PREMIRA® II microfibre highduster covers, 79,4% polyester/20,6% polypropylene
  • L×B:
    53,3×7,6 cm
  • Seq:
    01
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.