Objectives for REVOLVE 3M microscope
Plan achromat LWD; 0,4 NA, 20×, 2,8 mm
Katalog # ECHLJMAG0216
Leverandør: DISCOVER ECHO
Gå til:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Forstørrelse:20×
- Beskrivelse:Plan achromat LWD; 0,4 NA
- Seq:16
- Arbejdsafstand:2,8 mm