Earmuffs, LiteCom headsets, Peltor™
Product Details & Documents
Peltor™ LiteCom Headsets are hearing protectors with a built-in, pre-programmed two-way communication radio and an integrated noise cancelling microphone. They are built for demanding environments to help users communicate efficiently over hazardous noise.
- Hearing protector with built-in analogue two-way communication radio
- Eight channels in the PMR 446 MHz band
- 38 sub-channels
- Noise cancelling waterproof microphone provides voice-operated transmission (VOX) for hands-free use
- Voice guided menu system
- Auto power off function
- Clear battery status indication
- Communication radio in the PMR 446 MHz frequency range
Peltor™ LiteCom Headsets facilitate clear communicate while keeping user's hearing protected in noisy environments. These headsets have a built-in, pre-programmed two-way communication radio, which allows headset-to-headset conversation. Use eight channels in the PMR 446 MHz band, while 38 sub-channels let users communicate individually using the same channel. The noise cancelling microphone provides voice-operated transmission for hands-free use.
Documents
Specifikationer
- Farve:mørkeblå
- Beskrivelse:Peltor™ LiteCom two-way headset with J22 audio input, dark blue, neckband
- Seq:03
- Vægt:384 g
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



