Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Earmuffs, LiteCom headsets, Peltor™

Earmuffs, LiteCom headsets, Peltor™

Peltor™ LiteCom two-way headset with J22 audio input, dark blue, headband, mørkeblå, 418 g
Katalog # MMMAMT53H7P3E4400E
Leverandør:   3M
Earmuffs, LiteCom headsets, Peltor™
Katalog # MMMAMT53H7P3E4400E
Leverandør:  3M
Earmuffs, LiteCom headsets, Peltor™
Earmuffs, LiteCom headsets, Peltor™

Peltor™ LiteCom two-way headset with J22 audio input, dark blue, headband, mørkeblå, 418 g

Price
6.720,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Peltor™ LiteCom Headsets are hearing protectors with a built-in, pre-programmed two-way communication radio and an integrated noise cancelling microphone. They are built for demanding environments to help users communicate efficiently over hazardous noise.

  • Hearing protector with built-in analogue two-way communication radio
  • Eight channels in the PMR 446 MHz band
  • 38 sub-channels
  • Noise cancelling waterproof microphone provides voice-operated transmission (VOX) for hands-free use
  • Voice guided menu system
  • Auto power off function
  • Clear battery status indication
  • Communication radio in the PMR 446 MHz frequency range

Peltor™ LiteCom Headsets facilitate clear communicate while keeping user's hearing protected in noisy environments. These headsets have a built-in, pre-programmed two-way communication radio, which allows headset-to-headset conversation. Use eight channels in the PMR 446 MHz band, while 38 sub-channels let users communicate individually using the same channel. The noise cancelling microphone provides voice-operated transmission for hands-free use.

Specifikationer
  • Farve:
    mørkeblå
  • Beskrivelse:
    Peltor™ LiteCom two-way headset with J22 audio input, dark blue, headband
  • Seq:
    02
  • Vægt:
    418 g