High-Performance Chromatography Refrigerators, TSX
Product Details & Documents
The TSX high-performance chromatography refrigerators are designed for a variety of applications requiring close temperature control, full access to chromatography instrumentation, and easy set-up of instrumentation and apparatus within the chamber. The TSX chromatography series features industry-leading safety features to ensure maximum protection.
- Forced-air circulation for temperature uniformity at all shelf levels and Heat-free defrost for maximum temperature uniformity
- Quiet operation at just 52 dBA
- Two 50 mm side wall access ports with insulated hinged covers
- Four 50,8 mm casters for easy mobility; the front two are lockable
- Bright, LED interior lighting
- Keyed on/off switch with easy to use, set-point security
- Triple pane glass doors and self-closing door with 90° stop to assist with inventory loads
- Safety switch protected internal electrical outlet on all models (Duplex outleton A and D models, single outlet on V models)
- Energy Star certified and SNAP compliant
- DeviceLink compatible
TSX Series high-performance chromatography refrigerators-lab and clinical-grade storage that adapts to you and your environment. Temperature variation can have an impact on the viability and efficacy of vaccines, medication, reagents and other temperature-sensitive materials. These variations can shorten product shelf life, degrade enzymes and reduce vaccine effectiveness. Therefore, it is important to choose a high-performance refrigerator or freezer that has been designed for the demanding standards of the laboratory and clinical environment.
The sustainable, SNAP compliant choice - Thermo Scientific™ TSX Series high-performance refrigerators and freezers are designed with features that support sample protection, sustainability objectives and the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP). Our V-drive technology is designed to provide temperature uniformity that continually adapts to the lab or clinical environment, offering significant energy savings without compromising protection.
Whisper quiet operation - Compared to conventional refrigerant, high-performance models†, the TSX Series offers a whisper quiet operation, so you can bring the refrigerator or freezer out of the hallway and back into your lab without disrupting your audio environment.
Significant energy savings - While conventional refrigerant, high-performance refrigerators and freezers can run at very high kWh/day in energy usage, the TSX Series by comparison has been designed to reduce energy usage without compromising performance.
Note: Dimensions shown below are W×D×H.
Certifications: Meets cULus, CE, UPS 800, ISO Class 6, EPA Energy Star, EPA SNAP.
Specifikationer
- B×D×H udv.:1435×960×1994 mm
- Kapacitet:1447 L
- Beskrivelse:TSX high performance chromatography refrigerator CEE 7/7 plug
- Seq:03
- B×D×H indv.:1334×723×1473 mm
- Temperatur:2...8 °C
- Cat. No.:HERATSX5005CV
- Miljøcertificering:Energy Star; EPA SNAP Compliant
- Stik:CEE 7/7
- Elektriske specifikationer:208 - 230 V, 50/60 Hz
- Afrimning:Automatic
- Dørtype:2 glass swing doors
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



