Accessories for ASPEC® 241 System
Rack, code 345, holds 44 16×150 mm tubes
Catalog # GILI260440041
Supplier: GILSON LCMS
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Rack, code 345, holds 44 16×150 mm tubes
- Seq:19