Orders LinkContactUsLinkComponent
Accessories for ASPEC® 241 System
Accessories for ASPEC® 241 System
Rack, code 338, holds 64 12×32 mm vials (2 ml)
Katalog # GILI260440106
Leverandør:   GILSON LCMS
Accessories for ASPEC® 241 System
Katalog # GILI260440106
Leverandør:  GILSON LCMS
Accessories for ASPEC® 241 System
Accessories for ASPEC® 241 System
Rack, code 338, holds 64 12×32 mm vials (2 ml)
Price
3.030,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Rack, code 338, holds 64 12×32 mm vials (2 ml)
  • Seq:
    16