Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for ASPEC® 241 System

Accessories for ASPEC® 241 System

Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)
Catalog # GILI260440083
Supplier:   GILSON LCMS
Accessories for ASPEC® 241 System
Catalog # GILI260440083
Supplier:  GILSON LCMS
Accessories for ASPEC® 241 System
Accessories for ASPEC® 241 System

Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)

Price
3.030,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)
  • Seq:
    15