Accessories for PLC purification systems
Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)
Katalog # GILI150420
Leverandør: GILSON LCMS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)
- Seq:47
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



