Accessories for PLC purification systems
Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)
Katalog # GILI270433
Leverandør: GILSON LCMS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)
- Seq:36
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



