Accessories for PLC purification systems
Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)
Katalog # GILI150426
Leverandør: GILSON LCMS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)
- Seq:37