Accessories for PLC purification systems
Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204
Katalog # GILI150498
Leverandør: GILSON LCMS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204
- Seq:50