Accessories for PLC purification systems
Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
GILI152424
- Beskrivelse:Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
- Seq:39