Orders LinkContactUsLinkComponent
Accessories for PLC purification systems
Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
  GILI152424
Accessories for PLC purification systems
  GILI152424
Accessories for PLC purification systems
Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
7.200,00 kr
1 * 5 Items
  • Beskrivelse:
    Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
  • Seq:
    39