Accessories for PLC purification systems
Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)
Katalog # GILI150422
Leverandør: GILSON LCMS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)
- Seq:46



