Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for PLC purification systems

Accessories for PLC purification systems

Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)
Catalog # GILI152422
Supplier:   GILSON LCMS
Accessories for PLC purification systems
Catalog # GILI152422
Supplier:  GILSON LCMS
Accessories for PLC purification systems

Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)

Price
kr2,670.00
Log in for availability
1 * 5 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)
  • Seq:
    45