Orders LinkContactUsLinkComponent
Accessories for PLC purification systems
Accessories for PLC purification systems
Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
Katalog # GILI150425
Leverandør:   GILSON LCMS
Accessories for PLC purification systems
Katalog # GILI150425
Leverandør:  GILSON LCMS
Accessories for PLC purification systems
Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
Price
1.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
  • Seq:
    49