Orders LinkContactUsLinkComponent
Fraction collectors
Fraction collectors
Fraction collector FC203B with valve, HPLC purification technique, 110 - 120/220 - 240 V
Catalog # GILI171011DV
Supplier:   GILSON LCMS
Fraction collectors
Catalog # GILI171011DV
Supplier:  GILSON LCMS
Fraction collectors
Fraction collectors
Fraction collector FC203B with valve, HPLC purification technique, 110 - 120/220 - 240 V
Price
48.080,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone and software-controlled fraction collectors ranging from small footprint to large vessel capacity models. The built-in, easy-to-use keypad makes setup and operation simple and capable of interfacing with any HPLC system or control the system through your PC with TRILUTION® LC software.
  • Collect fractions by time, drop, or peak to suit all your LC applications
  • Choose the compact FC 203B for small-capacity collection or the FC 204 for large volumes and capacities
  • In peak mode, collectors monitor a detector signal and identify peaks using either an adaptive-slope algorithm or a specified millivolt threshold
  • Time: Up to 10 programmable time windows can be added for collecting only what you want, while discarding the column’s void volume, peaks of no interest, and equilibration volumes
  • 3-way diverter valve prevents contamination of collected fractions (optional)
  • Compact size fits in most fume hoods
FC203B and FC203B with valve: choose among peak, drop, or time collection options with the FC 203B fraction collector. In peak mode, the FC 203B monitors a detector signal and identifies peaks using either an adaptive-slope algorithm or a specified millivolt threshold. With up to ten programmable time windows to use, collect only what you want and discard the column void volume, peaks of no interest, and equilibration volumes. Multicolumn adapters are available for collecting up to 18 separate channels and an optional 3-way diverter valve prevents contamination of collected fractions.
FC204: Collect samples by peak, drop, or time using the FC 204 Fraction Collector. Add up to ten programmable time windows to collect only what you want, eliminating void volumes, equilibration volumes, and peaks of no interest. In peak mode, the collector monitors a detector signal, identifying peaks using an adaptive-slope algorithm or a specified millivolt threshold. The 3-way diverter valve prevents contamination of fractions. Multicolumn adapters allow for collection from up to 18 separate channels.
Specifications
  • Beskrivelse:
    Fraction collector FC203B with valve, HPLC purification technique, 110 - 120/220 - 240 V
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150427
    Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (20 ml)
    Log in for availability
    1 * 1 Items - 687,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150420
    Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)
    Log in for availability
    1 * 1 Items - 679,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150422
    Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)
    Log in for availability
    1 * 1 Items - 916,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150424
    Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml), for use with FC204
    Log in for availability
    1 * 1 Items - 1.500,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150425
    Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
    Log in for availability
    1 * 1 Items - 1.060,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150426
    Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)
    Log in for availability
    1 * 1 Items - 687,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150429
    Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
    Log in for availability
    1 * 1 Items - 996,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI150498
    Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204
    Log in for availability
    1 * 1 Items - 2.670,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI152420
    Rack, code 28, holds quantity of 108, 10×75 mm tubes (5/pack)
    Log in for availability
    1 * 5 Items - 3.030,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI152422
    Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)
    Log in for availability
    1 * 5 Items - 2.670,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI152424
    Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
    Log in for availability
    1 * 5 Items - 7.200,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI152426
    Rack, code 23, holds quantity of 44, 17×55 scintilation vials (5/pack)
    Log in for availability
    1 * 5 Items - 3.210,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI152427
    Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (5/pack)
    Log in for availability
    1 * 5 Items - 3.180,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI270433
    Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)
    Log in for availability
    1 * 1 Items - 2.110,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170413
    Rack, code 11, holds quantity of 105, 13×100 mm, 9 ml, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 9.120,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI2704342
    Rack, code 29LE, holds quantity of 60, 1,5 ml microcentrifuge tubes
    Log in for availability
    1 * 1 Items - 2.620,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI2704341
    Rack, code 29SE, holds quantity of 60, 0,5 ml microcentrifuge tubes
    Log in for availability
    1 * 1 Items - 3.200,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI130412
    Rack, code 4, holds quantity of 80, 12×75 mm tubes, 5 ml, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 5.090,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170414
    Rack, code 14, holds quantity of 128, 12×75 or 100 mm, 5 ml, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 7.240,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI270430
    Rack, code 0, holds quantity of 80, 12×32 mm vials, 2 ml, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 518,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170730
    3-way valve kit with 1,5 mm Int.Ø, FC203, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 6.700,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170735
    FC-204 Prep valve assembly package, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 12.140,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170415
    Rack, code 15, holds 1×96-well microplate, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 851,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170416
    Rack, code 16, holds 4×96-well microplates, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 1.800,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170417
    Rack heightener, code 17, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 1.650,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170418
    Rack, code 17, microplate, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 3.170,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170734
    3-way valve kit with 1,5 mm Int.Ø, FC204, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 7.460,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI170416D
    Rack, code 16D, holds 4 deep well microplates, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 1.800,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • GILI174732
    Multiple column adapter, for use with fraction collectors
    Log in for availability
    1 * 1 Items - 6.610,00 kr