Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

2% Potassium (K) in 1% HCl 1%, Plastic bottle, 100 mL
Katalog # HONCE4302-100ML
Leverandør:   Honeywell Chemicals
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Katalog # HONCE4302-100ML
Leverandør:  Honeywell Chemicals
CAS nummer:  
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

2% Potassium (K) in 1% HCl 1%, Plastic bottle, 100 mL

CAS nummer:  
Price
2.000,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)

Leveringsinformation: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.

Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 mL
  • Beskrivelse:
    2% Potassium (K) in 1% HCl 1%

Specifikationer