Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
2% Lithium (Li) in 1% HNO₃, Plastic bottle, 100 mL
Katalog # HONCE4301-100ML
Leverandør: Honeywell Chemicals
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Katalog # HONCE4301-100ML
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Leveringsinformation: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 mL
- Beskrivelse:2% Lithium (Li) in 1% HNO₃
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



