Titanium AAS Standards, (CRMs), Fluka™
Titanium, Ti, 1000 mg/L, 5% HNO₃/<0.5% HF, Plastic bottle, 500 mL
Catalog # HONCE4077-500ML
Supplier: Honeywell Chemicals
Titanium AAS Standards, (CRMs), Fluka™
Catalog # HONCE4077-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Koncentration:1000 mg/L
- Matrice:5% HNO₃/<0.5% HF
- Pakning:Plastic bottle
- Pakn:500 mL
- Element:Titanium
- Symbol:Ti



