Titanium AAS Standards, (CRMs), Fluka™
Titanium, Ti, 1000 mg/L, 5% HNO₃/<0.5% HF, Plastic bottle, 100 mL
Katalog # HONCE4077-100ML
Leverandør: Honeywell Chemicals
Titanium AAS Standards, (CRMs), Fluka™
Katalog # HONCE4077-100ML
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifikationer
Specifikationer
- Koncentration:1000 mg/L
- Matrice:5% HNO₃/<0.5% HF
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Titanium
- Symbol:Ti



