Selenium(IV) ICP Standards, (CRMs), Fluka™
Selenium IV, Se, 1000 mg/L, 2% HNO₃, Plastic bottle, 100 mL
Katalog # HONCE1507-100ML
Leverandør: Honeywell Chemicals
Selenium(IV) ICP Standards, (CRMs), Fluka™
Katalog # HONCE1507-100ML
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifikationer
Specifikationer
- Koncentration:1000 mg/L
- Matrice:2% HNO₃
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Selenium IV
- Symbol:Se
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



