Selenium(IV) ICP Standards, (CRMs), Fluka™
Selenium IV, Se, 100 mg/L, 2% HNO₃, Plastic bottle, 100 mL
Katalog # HONCE1503-100ML
Leverandør: Honeywell Chemicals
Selenium(IV) ICP Standards, (CRMs), Fluka™
Katalog # HONCE1503-100ML
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifikationer
Specifikationer
- Koncentration:100 mg/L
- Matrice:2% HNO₃
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Selenium IV
- Symbol:Se



