Lead ICP standards, (CRMs), Fluka™
Bly, Pb, 10000 mg/L, 5% HNO₃, Plastic bottle, 100 mL
HONCE1170-100ML
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
- Koncentration:10000 mg/L
- Matrice:5% HNO₃
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Bly
- Symbol:Pb
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



