Orders LinkContactUsLinkComponent
ICP standard solutions, Fluka™
ICP standard solutions, Fluka™
ICP Standard (15 Components), 1 - 200 mg/L, 5% HNO₃, 100 mL
Katalog # HONCE2213-100ML
Leverandør:   Honeywell Chemicals
ICP standard solutions, Fluka™
Katalog # HONCE2213-100ML
Leverandør:  Honeywell Chemicals
CAS nummer:  
ICP standard solutions, Fluka™
ICP Standard (15 Components), 1 - 200 mg/L, 5% HNO₃, 100 mL
CAS nummer:  
Price
3.600,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Certified Reference Materials (CRMs).
  • Traceable to NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Specifikationer
Specifikationer
  • Koncentration:
    1 - 200 mg/L
  • Matrice:
    5% HNO₃
  • Pakn:
    100 mL
  • Beskrivelse:
    ICP Standard (15 Components)