Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 5 supplement

HyClone Cell Boost™ 5 supplement

Cell Boost™ 5, Liquid, 1000 ml, 1.000 ml
Katalog # SH31117.02
HyClone Cell Boost™ 5 supplement
Katalog # SH31117.02
CAS nummer:  
HyClone Cell Boost™ 5 supplement
HyClone Cell Boost™ 5 supplement

Cell Boost™ 5, Liquid, 1000 ml, 1.000 ml

CAS nummer:  
Price
1.920,00 kr
Log ind for tilgængelighed
1 * 1.000 mL
Product Details & Documents

Supplements cell culture with lipids, amino acids, vitamins, and growth factors. Manufactured to meet cGMP manufacturing standards and QC specifications.

  • Designed to support modern manufacturing processes, Cell Boost 5 is chemically defined (CD), animal-derived component-free (ADCF), protein-free, and hydrolysate-free
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 5 Supplement provides lipids, amino acids, vitamins, and growth factors to support fed-batch cultures in research and bioprocessing applications. It is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

This HyClone cell culture supplement has been developed for recombinant protein production with various cell lines including Chinese hamster ovary (CHO) cell line, human embryonic kidney (HEK) 293, NS0, and hybridoma cells.

Recommended supplementation range is between 20 to 200 ml/L during mid to late growth phase. It is anticipated that this will be day 3 to 4 of the culture. It is suggested that L-glutamine is monitored separately in those cultures where it is present in the culture medium and supplemented as needed.

Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.

Specifikationer

Specifikationer

  • Pakn:
    1.000 ml
  • Beskrivelse:
    Cell Boost™ 5
  • Størrelse:
    1000 ml
  • Form:
    Liquid
  • Cat. No.:
    SH31117.02
  • Opbevaringstemperatur:
    2…8 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.