Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 7b supplement

HyClone Cell Boost™ 7b supplement

Cell Boost™ 7b, Liquid, 100 ml, 100 ml
Katalog # SH31120.01
HyClone Cell Boost™ 7b supplement
Katalog # SH31120.01
CAS nummer:  
HyClone Cell Boost™ 7b supplement
HyClone Cell Boost™ 7b supplement

Cell Boost™ 7b, Liquid, 100 ml, 100 ml

CAS nummer:  
Price
635,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.

  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pHSupplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
  • Non sterile

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines. Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

Specifikationer

Specifikationer

  • Pakn:
    100 ml
  • Beskrivelse:
    Cell Boost™ 7b
  • Størrelse:
    100 ml
  • Form:
    Liquid
  • Cat. No.:
    SH31120.01
  • Shelf life:
    18 months
  • Opbevaringstemperatur:
    2…8 °C
  • Opbevaring betingelser:
    Away from light and moisture

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.