Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 7a supplement

HyClone Cell Boost™ 7a supplement

Cell Boost™ 7a, Liquid, 500 ml, 500 ml
Katalog # SH31119.01
HyClone Cell Boost™ 7a supplement
Katalog # SH31119.01
CAS nummer:  
HyClone Cell Boost™ 7a supplement
HyClone Cell Boost™ 7a supplement

Cell Boost™ 7a, Liquid, 500 ml, 500 ml

CAS nummer:  
Price
1.560,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose. Manufactured to meet cGMP manufacturing standards and QC specifications.

  • Designed to support modern manufacturing processes, Cell Boost 7a is chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines. Cell Boost 7a is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

This HyClone cell culture supplement has been developed for high-yield recombinant protein production when using various Chinese hamster ovary (CHO) cell lines.

Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.

Specifikationer

Specifikationer

  • Pakn:
    500 ml
  • Beskrivelse:
    Cell Boost™ 7a
  • Størrelse:
    500 ml
  • Form:
    Liquid
  • Sammensætning:
    Without: L-glutamine <br />With: Poloxamer 188
  • Cat. No.:
    SH31119.01
  • Shelf life:
    18 months
  • Opbevaringstemperatur:
    2…8 °C
  • Opbevaring betingelser:
    Away from light and moisture

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.