Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 39,37 - 54,61 mm tall, 690×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
Katalog # MATR1950839
Leverandør: THERMO MATRIX TECHNOLOGIES
Accessories for CryoPlus Sample Storage Systems
Katalog # MATR1950839
Leverandør: THERMO MATRIX TECHNOLOGIES
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack for Matrix 2D plates measuring 39,37 - 54,61 mm tall, 690×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
- Seq:11