Transport medium, Mediatech (Corning)
Transport medium, 500 ml, 6
Leverandør: Corning
Gå til:
Product Details & Documents
Corning® transport medium is formulated using the final ingredient concentrations outlined in CDC's SOP#: DSR-052-01 'Preparation of viral transport medium'.
- cGMP grade
- Aseptically sterile-filtered
- 10⁻³ SAL, labelled 'Sterile A'
Transport medium is an isotonic solution at neutral pH containing protective proteins, antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi.
Documents
Specifikationer
Specifikationer
- Pakn:6
- Beskrivelse:Transport medium, 500 ml