Orders LinkContactUsLinkComponent
Transport medium, Mediatech (Corning)
Transport medium, Mediatech (Corning)
Transport medium, 500 ml, 6
Katalog # MDTC25-500-CV
Leverandør:   Corning
Transport medium, Mediatech (Corning)
Katalog # MDTC25-500-CV
Leverandør:  Corning
CAS nummer:  
Transport medium, Mediatech (Corning)
Transport medium, Mediatech (Corning)
Transport medium, 500 ml, 6
CAS nummer:  
Price
2.890,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
Corning® transport medium is formulated using the final ingredient concentrations outlined in CDC's SOP#: DSR-052-01 'Preparation of viral transport medium'.
  • cGMP grade
  • Aseptically sterile-filtered
  • 10⁻³ SAL, labelled 'Sterile A'
Transport medium is an isotonic solution at neutral pH containing protective proteins, antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi.
Documents
Specifikationer
Specifikationer
  • Pakn:
    6
  • Beskrivelse:
    Transport medium, 500 ml