Avantor® Hichrom HI-5 Basic, GC Columns
GC column, Kapillær, 1,00 µm, 0,32 mm, 15 m
HI03-32-100-15
: Avantor
Max temperature limits may change depending on the film thickness.
- Kolonne form:Kapillær
- Filmtykkelse:1,00 µm
- Ø indv.:0,32 mm
- Beskrivelse:GC column
- Længde:15 m
- Cat. No.:HI03-32-100-15
- Crossbond:Yes
- Phase composition:5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- Polarity:Low-polarity stationary phase
- Phase name:HI-5 Basic
Accessories
- Beskrivelse
- HI0-100-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 100 Items - 6.330,00 kr
- Beskrivelse
- HI0-10-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 10 Items - 867,00 kr
- Beskrivelse
- HI0-10-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 10 Items - 7.540,00 kr
- Beskrivelse
- HI0-1-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 1 Items - 780,00 kr
- Beskrivelse
- HI0-25-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 25 Items - 1.690,00 kr
- Beskrivelse
- HI0-2-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 2 Items - 1.560,00 kr
- Beskrivelse
- HI0-50-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 50 Items - 3.250,00 kr
- Beskrivelse
- HI0-5-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 5 Items - 582,00 kr
- Beskrivelse
- HI0-5-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm1 * 5 Items - 3.860,00 kr