Avantor® Hichrom HI-5 Basic, GC Columns
GC column, Kapillær, 1,00 µm, 0,25 mm, 15 m
Katalog # HI03-25-100-15
Leverandør: Avantor
Gå til:
Product Details & Documents
Max temperature limits may change depending on the film thickness.
Specifikationer
- Kolonne form:Kapillær
- Filmtykkelse:1,00 µm
- Ø indv.:0,25 mm
- Beskrivelse:GC column
- Længde:15 m
- Cat. No.:HI03-25-100-15
- Crossbond:Yes
- Phase composition:5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- Polarity:Low-polarity stationary phase
- Phase name:HI-5 Basic
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-100-PF-CLog ind for tilgængelighed1 * 100 Items - 6.330,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PF-CLog ind for tilgængelighed1 * 10 Items - 867,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PFY-CLog ind for tilgængelighed1 * 10 Items - 7.540,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-1-PFY-CLog ind for tilgængelighed1 * 1 Items - 780,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-25-PF-CLog ind for tilgængelighed1 * 25 Items - 1.690,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-2-PFY-CLog ind for tilgængelighed1 * 2 Items - 1.560,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-50-PF-CLog ind for tilgængelighed1 * 50 Items - 3.250,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PF-CLog ind for tilgængelighed1 * 5 Items - 582,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PFY-CLog ind for tilgængelighed1 * 5 Items - 3.860,00 kr