Screw Caps for Bottles
Screw cap, tamper evident, with PTFE seal, 28 mm, hvid, HDPE
Katalog # RIXI8-1316-28-WS
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Specifikationer
- Gevind:28 mm
- Farve:hvid
- Beskrivelse:Screw cap, tamper evident, with PTFE seal
- Materiale:HDPE
- Seq:29