Re-Usable Cleaning Coupons
304 stainless steel - satin, 50×50 mm
Katalog # 300-0611
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
- All popular sizes available
- Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifikationer
- Beskrivelse:304 stainless steel - satin
- L×B:50×50 mm
- Seq:03