Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Micro-spatula samplers, SteriWare

Micro-spatula samplers, SteriWare

Sterile micro-spatula, 13 mm blade width, PP, blå, 2 ml
Katalog # 300-0491
Leverandør:   Sampling Systems
Micro-spatula samplers, SteriWare
Katalog # 300-0491
Leverandør:  Sampling Systems
Micro-spatula samplers, SteriWare
Micro-spatula samplers, SteriWare

Sterile micro-spatula, 13 mm blade width, PP, blå, 2 ml

Price
1.360,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.

  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use

Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.

Specifikationer
  • Volumen:
    2 ml
  • Farve:
    blå
  • Beskrivelse:
    Sterile micro-spatula, 13 mm blade width
  • Materiale:
    PP
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.