Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Micro-spatula samplers, SteriWare

Sterile micro-spatula, 4 mm blade width, PP, blå, 0,3 ml
  300-0489
Micro-spatula samplers, SteriWare
  300-0489
Micro-spatula samplers, SteriWare
Micro-spatula samplers, SteriWare

Sterile micro-spatula, 4 mm blade width, PP, blå, 0,3 ml

 
1.680,00 kr
   
1 * 100 Items
 

Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.

  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use

Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.

 
  • Volumen:
    0,3 ml
  • Farve:
    blå
  • Beskrivelse:
    Sterile micro-spatula, 4 mm blade width
  • Materiale:
    PP
  • Seq:
    01