Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Transport medium, Mediatech (Corning)

Transport medium, Mediatech (Corning)

Transport medium, 100 ml, 12 Months, 6
Catalog # MDTC25-500-CI
Supplier:   Corning
Transport medium, Mediatech (Corning)
Catalog # MDTC25-500-CI
Supplier:  Corning
CAS Number:  
Transport medium, Mediatech (Corning)
Transport medium, Mediatech (Corning)

Transport medium, 100 ml, 12 Months, 6

CAS Number:  
Price
1.140,00 kr
Log in for availability
1 * 6 Items
Product Details & Documents

Corning® transport medium is formulated using the final ingredient concentrations outlined in CDC's SOP#: DSR-052-01 'Preparation of viral transport medium'.

  • cGMP grade
  • Aseptically sterile-filtered
  • 10⁻³ SAL, labelled 'Sterile A'

Transport medium is an isotonic solution at neutral pH containing protective proteins, antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi.

Documents

Specifications

Specifications

  • Pakn:
    6
  • Beskrivelse:
    Transport medium, 100 ml
  • Holdbarhed:
    12 Months
  • Cat. No.:
    MDTC25-500-CI
  • Opbevaring betingelser:
    2...8 °C

Specifications