Orders LinkContactUsLinkComponent
Computers for Laboratory Equipment
PC Dell Optiplex XE4 Tower - Processor Intel Core i5-12500 (6C - Up to 4,60 GHz); 8 GB DDR4 Memory - 512 GB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 10 IoT Ent. 2021 LTSC Value license operating system
  906-0235
Computers for Laboratory Equipment
  906-0235
Computers for Laboratory Equipment
Computers for Laboratory Equipment
PC Dell Optiplex XE4 Tower - Processor Intel Core i5-12500 (6C - Up to 4,60 GHz); 8 GB DDR4 Memory - 512 GB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 10 IoT Ent. 2021 LTSC Value license operating system
13.320,00 kr
1 * 1 Items
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
  • Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
  • Compact footprint with integrated PC-monitor solutions
  • Pre-installed language packs
  • Keyboards with all common European language layouts
  • Compatible with main CDS (Chromatography Data Systems)
  • Beskrivelse:
    PC Dell Optiplex XE4 Tower - Processor Intel Core i5-12500 (6C - Up to 4,60 GHz); 8 GB DDR4 Memory - 512 GB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 10 IoT Ent. 2021 LTSC Value license operating system
  • Seq:
    05
Complementary Products
  •  
    Beskrivelse
  • 906-0219
    Dell™ mouse, bundled with Dell™ keyboard, DE
    1 * 1 Items - 231,00 kr
  • 906-0211
    Dell™ mouse, bundled with Dell™ keyboard, FR
    1 * 1 Items - 231,00 kr