0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
[EN]SODIUM HYDROXIDE SOLUTION 10 MOL/L E 1 * 1 L
[EN]SODIUM HYDROXIDE SOLUTION 10 MOL/L E 1 * 1 L
4.80648.1002
:
MERCK PRODUCTION CHEMICALS
[EN]SODIUM HYDROXIDE SOLUTION 10 MOL/L E 1 * 1 L
4.80648.1002
:
MERCK PRODUCTION CHEMICALS
[EN]SODIUM HYDROXIDE SOLUTION 10 MOL/L E 1 * 1 L
1.050,00 kr
1 * 1 L
-
+
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
#: 20060.320
: VWR Chemicals
Handsker, nitril, Kimtech™ Purple Nitrile™
#: 112-4847
: Ansell