LED modules
LED module 6 V/3 W 5000 K +/-300 K for BA 210 Elite
Catalog # MOTI1101002405631
Supplier: MOTIC
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:LED module 6 V/3 W 5000 K +/-300 K for BA 210 Elite
- Seq:04