Cleaning cloths, Microfibre Light
hvid, 340×400 mm, 40 sheets per pack
Leverandør: PGI NONWOVEN
Gå til:
Product Details & Documents
Microfibre Light cloths provide the same superior cleaning performance as traditional woven microfibre cloths without the risk of cross contamination. Designed for short term use.
- Removes 99% of bacteria from surfaces, improving hygiene
- Can be disposed of after use, reducing time spent by cleaning staff on non value added tasks
- Unique structure of the Microfibre Light cloth ensures that bacteria are trapped in the cloth and not released onto the next surface
- Can be used in food applications (FCC certificate)
Ideal for healthcare applications, food processing industries, building care and janitorial tasks.
Specifikationer
- Pakket:40 sheets per pack
- Farve:hvid
- L×B:340×400 mm