Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Rotigotine Related Compounds, Reference Standard

Rotigotine Related Compound C, USP reference standard, Vial, 25 mg
Katalog # 1606117.
Leverandør:   USP
Rotigotine Related Compounds, Reference Standard
Katalog # 1606117.
Leverandør:  USP
CAS nummer:  
Rotigotine Related Compounds, Reference Standard

Rotigotine Related Compound C, USP reference standard, Vial, 25 mg

CAS nummer:  
Price
11.280,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Pakn:
    25 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Rotigotine Related Compound C
  • Cat. No.:
    1606117.
  • Beskrivelse:
    Rotigotine Related Compound C
  • Synonym(s):
    (S)-6-{[2-(Thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol hydrochloride

Specifikationer