Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Nicotine Related Compounds, Reference Standard

Nicotine Related Compounds, Reference Standard

Nicotine Related Compound D, USP reference standard, Vial, 20 mg
Katalog # 1463359.
Leverandør:   USP
Nicotine Related Compounds, Reference Standard
Katalog # 1463359.
Leverandør:  USP
CAS nummer:  
Nicotine Related Compounds, Reference Standard

Nicotine Related Compound D, USP reference standard, Vial, 20 mg

CAS nummer:  
Price
kr9,770.00
Log ind for tilgængelighed
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Pakn:
    20 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Nicotine Related Compound D
  • Cat. No.:
    1463359.
  • Beskrivelse:
    Nicotine Related Compound D
  • Synonym(s):
    3-(4,5-dihydro-3H-pyrrol-2-yl)pyridine fumarate

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.