Orders LinkContactUsLinkComponent
Deferasirox, Reference Standards
Deferasirox, Reference Standards
Deferasirox System Suitability Mixture, USP reference standard, Vial, 20 mg
Katalog # 1165667.
Leverandør:   USP
Deferasirox, Reference Standards
Katalog # 1165667.
Leverandør:  USP
CAS nummer:  
Deferasirox, Reference Standards
Deferasirox System Suitability Mixture, USP reference standard, Vial, 20 mg
CAS nummer:  
Price
9.070,00 kr
Log ind for tilgængelighed
1 * 20 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Deferasirox System Suitability Mixture: A mixture containing Deferasirox, Benzoxazinone analog, Deferasirox ethyl ester, Deferasirox 1,2-isomer, Deferasirox 1,3-isomer and Salicylimide
Certifications: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
  • Pakning:
    Vial
  • Pakn:
    20 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Deferasirox System Suitability Mixture
  • UN No.:
    3077
  • Cat. No.:
    1165667.
  • Beskrivelse:
    Deferasirox